SP Bel-Art Cleanware™ Autoclavable Waste Bags, Bel-Art Products, a part of SP
# de catalogue 11215-853
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- W×L:203×254 mm (8×10")
- Cat. no.:11215-853
- Supplier No.:131741008
Spécifications
A propos de cet article
Disposable waste bags free bench or hood space when hung from walls, cabinets, or virtually anywhere.
Disposable waste bags free bench or hood space when hung from walls, cabinets, or virtually anywhere.
Simply peel off the backing on the adhesive strip across the top and hang close to your work area. The back panel is longer than the front for easy opening. Made of 1 mil thick, high-molecular weight, high-density (HMHD) polyethylene, which is steam autoclavable at 121 °C (250 °F). Conforms to ASTM 1922 Tear Resistance and ASTM 1709 Dart Impact Tests. Maximum weight of contents should not exceed 1.4 kg (3 lbs.).
Customers Who Bought This Also Bought