ContactUsLinkComponent
VWR* Double Thick Polyethylene Biohazard Bags
VWR® Autoclavable PE Biohazard Bags, 4 mil
Catalog # 11215-826
Supplier:  VWR International
Product available on GSA Advantage®
Small Business Enterprise
VWR* Double Thick Polyethylene Biohazard Bags
VWR® Autoclavable PE Biohazard Bags, 4 mil
Catalog # 11215-826
Supplier:  VWR International
Product available on GSA Advantage®
Small Business Enterprise
Specifications
  • Color:
    red
  • Cat. no.:
    11215-826
  • W×L:
    610×914 mm (24×36")
Specifications
About this item
These heavy-duty bags with extra puncture resistance and high impact strength are constructed of high-molecular, high-density 4 mm polyethylene plastic.
  • Bright red color for quick identification
  • Clearly labeled with biohazard warnings
Autoclavable up to 121 °C (250 °F). Ideal for sterilizing animal cages. Each package includes 20.3 cm (8") twist ties. Bags meet many state and local regulations.
Customers Who Bought This Also Bought