Order Entry
ContactUsLinkComponent
Heavy-Weight Bags
SP Bel-Art Utility Bags, Bel-Art Products, a part of SP
# de catalogue 11215-359
Heavy-Weight Bags
SP Bel-Art Utility Bags, Bel-Art Products, a part of SP
# de catalogue 11215-359

Spécifications

  • W×L:
    254×457 mm (10×18")
  • Thickness:
    4 mil
  • Cat. no.:
    11215-359
  • Supplier No.:
    131781018

Spécifications

A propos de cet article

These all-purpose polyethylene bags are available in two weights.

  • Polyethylene construction
  • Latex-free
  • Not autoclavable
  • Reusable and transparent

They are ideal for shipping or storage and are especially useful as sample bags.

Certifications: FDA approved material.
Customers Who Bought This Also Bought