VWR® Wafer Box Sealing Tape, PE
# de catalogue 11211-910
Fournisseur: VWR International
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Width:5.1 cm (2")
- Color:Transparent
Spécifications
A propos de cet article
Ideal for packaging and transporting of wafer boxes, this low-density polyethylene tape is low in halogens and leachable chlorides.
- Metal free adhesive
- Polyethylene film fabrication
- Acrylic adhesive minimizes outgassing
- Excellent chemical resistance
- Good conformability
Polyethylene film and acrylic adhesive minimize outgassing and offer good 'clean peel' properties. Offers excellent chemical resistance and good conformability.
Customers Who Bought This Also Bought