Axygen® Pipette Filter Tip, Axygen®, Corning
Catalog # 10011-882
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Volume:20 µl
- Cat. no.:10011-882
- Sterile:-
- Supplier No.:TF-20
Specifications
About this item
Axygen brand filter tips allow free passage of air while preventing passage of aerosols.
The filters contain no leachable additives.
Customers Who Bought This Also Bought