ContactUsLinkComponent
Filter Pipet Tip
Axygen® Pipette Filter Tip, Axygen®, Corning
Catalog # 10011-882
Supplier:  Corning
Product available on GSA Advantage®
Filter Pipet Tip
Axygen® Pipette Filter Tip, Axygen®, Corning
Catalog # 10011-882
Supplier:  Corning
Product available on GSA Advantage®
Specifications
  • Volume:
    20 µl
  • Cat. no.:
    10011-882
  • Sterile:
    -
  • Supplier No.:
    TF-20
Specifications
About this item
Axygen brand filter tips allow free passage of air while preventing passage of aerosols.
The filters contain no leachable additives.
Customers Who Bought This Also Bought