Axygen® Pipette Filter Tip, Axygen®, Corning
# de catalogue 10011-878
Fournisseur: Corning
Product available on GSA Advantage®
Spécifications
- Volume:100 µl
- Cat. no.:10011-878
- Sterile:-
- Supplier No.:TF-100
Spécifications
A propos de cet article
Axygen brand filter tips allow free passage of air while preventing passage of aerosols.
The filters contain no leachable additives.
Customers Who Bought This Also Bought