ContactUsLinkComponent
Presterilized Microtube
Axygen® Snaplock Microcentrifuge Tubes, 0.6/1.5/1.7/2.0/5.0 ml, Corning
Catalog # 10011-724
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
undefined
Axygen® Snaplock Microcentrifuge Tubes, 0.6/1.5/1.7/2.0/5.0 ml, Corning
Catalog # 10011-724
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
Specifications
  • Sterility:
    Sterile
  • Volume:
    1.7 ml
  • Color:
    Clear
  • Description:
    Microtube
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Maximum RCF:
    20000×g
  • Supplier No.:
    MCT-175-C-S
Specifications
About this item
High quality PP, graduated.
  • Maxymum Recovery® microtubes with ultra-smooth surfaces prevent adhesion of proteins and nucleic acids after long periods of storage
  • Certified free from DNase, RNase and endotoxin
Microtubes made from PP homopolymer for excellent sample visibility. Available in an ultra-transparent (MaxyClear) version clear or in assorted colours, with frosted writing area.
Frequently Bought Together