Axygen® Self-Standing Screw Top Microcentrifuge Tubes with Caps, Corning
Catalog # 10011-566
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Sterility:Sterile
- Volume:1.5 ml
- Description:Microtube
- Cat. no.:10011-566
- Maximum RCF:20000×g
- Cap Color:Clear
- Supplier No.:SCT-150-SS-C-S
Specifications
About this item
Microcentrifuge tubes serve a broad range of laboratory needs including collection, centrifugation, long-term storage, and OEM packaging and shipping of restriction enzymes or other DNA/RNA modifying reagents.
Threaded O-ring design ensures a uniform closure. Ribs on cap and tube are designed to fit the AxyRack™ microtube racks (see 10011-272 series) to allow single-handed opening and closing. MAXYMum Recovery™ tubes feature an ultra-smooth surface completely free of occlusions and cavities, virtually eliminating sample retention while improving accuracy. Certified RNase-, DNase-, and pyrogen-free.
Customers Who Bought This Also Bought