Spécifications
- Sterility:Sterile
- Volume:1.5 ml
- Maximum RCF:20000×g
- Cap Color:Clear
- Supplier No.:SCT-150-C-S
Spécifications
A propos de cet article
These conical-bottom screw cap microcentrifuge tubes serve a broad range of laboratory needs including collection, pressing, centrifugation, long-term storage, and OEM packaging and shipping of restriction enzymes or other DNA/RNA modifying reagents.
These conical-bottom screw cap microcentrifuge tubes serve a broad range of laboratory needs including collection, pressing, centrifugation, long-term storage, and OEM packaging and shipping of restriction enzymes or other DNA/RNA modifying reagents.
Color-coded caps provide a syringe port that is easily pierceable for radioisotope labeling and fraction collecting. The threaded O-ring design ensures a uniform closure allowing centrifugation up to 20000×g. Microcentrifuge tubes have ribs on the cap and tube to fit the Axyrack™ Microtube Racks to allow single-handed opening and closing. Polypropylene tubes are certified RNase-, DNase-, and pyrogen-free.
All microtubes include non-looped caps with O-rings. Presterilized microtubes are supplied with cap attached.