ContactUsLinkComponent
Universal Fit Pipet Tip
Axygen® Research-Grade Bevelled Reference Pipette Tips 200 µl, Corning
Catalog # 10011-376
Supplier:  Corning
Product available on GSA Advantage®
Universal Fit Pipet Tip
Axygen® Research-Grade Bevelled Reference Pipette Tips 200 µl, Corning
Catalog # 10011-376
Supplier:  Corning
Product available on GSA Advantage®
Specifications
  • Volume:
    200 µl
  • Tip Color:
    Yellow
  • Packed:
    Racked
  • Cat. no.:
    10011-376
  • Supplier No.:
    TR-222-Y-R
Specifications
About this item
Axygen tips are designed to fit a wide variety of single-channel and multichannel pipettors.
Flexible walls and a series of internal sealing rings ensure a secure fit and less force required to load and eject. Accuracy and precision testing is performed for every manufacturing lot using the finest pipettors in the industry.
Customers Who Bought This Also Bought