ContactUsLinkComponent
Orion Gel-Filled Triode
Orion™ Redox/ORP/Temp Electrodes, Thermo Scientific
# de catalogue 10010-736
Orion Gel-Filled Triode
Orion™ Redox/ORP/Temp Electrodes, Thermo Scientific
# de catalogue 10010-736
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Gel-Filled Redox/ORP/ATC Electrode
  • Cat. no.:
    10010-736
  • Supplier No.:
    9179BNMD
Spécifications
A propos de cet article
Efficiently measure ORP and temperature simultaneously without a separate reference electrode in a single instance
  • Available in epoxy triode low maintenance and epoxy refillable triode construction
  • Built-in thermistor for Automatic Temperature Compensation (ATC)
  • Not designed for amperometric or constant current titrations, such as Karl Fischer analysis
Orion™ Redox/ORP/Temp Electrodes measure ORP and temperature using a platinum redox sensor, a silver/silver chloride internal reference system, and a built-in thermistor which is adapted for Automatic Temperature Compensation (ATC).
Customers Who Bought This Also Bought