VANES
# de catalogue WT41-1059
Fournisseur: Welch Vacuum
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Fournisseur: VWR International
$881.80 to $2,807.56
Chloroform ≥99.90% stabilized, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Fournisseur: VWR International
$438.39 to $1,639.42
Methanol ≥99.8%, GR ACS, meets analytical specification for testing USP/NF monographs, Supelco®
Fournisseur: MilliporeSigma
$1,413.13
Sodium sulfate, anhydrous ≥99.0%, granular, Tracepur® 10-60 mesh, Supelco®
Fournisseur: MilliporeSigma
$1,352.49