Sodium hydroxide 0.0100 N (N/100)
# de catalogue RC72951
Fournisseur: Ricca Chemical
Small Business Enterprise
Numéro CAS: 1310-73-2
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Poly
- Size:1 gal.
- Cat. no.:RC72951
- Supplier No.:R7295000-4A
Spécifications
Résultats des tests de spécification
- Appearance:Clear, colorless, odorless
- Assay at 20 °C (traceable to NIST SRM 723):0.01000 ± 0.00010 N at 20.0 °C
- Sodium hydroxide 0.0100 N (N/100):
Résultats des tests de spécification
A propos de cet article
Synonymes: Caustic soda
Densité:1 g/cm³ (25 °C)
ADR:8,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Pt ébul.:|Point d'ébullition:
:Pt ébul.: 1390 °C (1013 hPa)
:Pt Fusion:|Point de fusion:
:Pt Fusion: 323 °C
:Pt Eclair:|Point eclaire:
:M.W. 40 g/mol
Température d'entreposage:Température ambiante
Température d'entreposage:Ambient
EINECS:215-185-5
:323
:1390
ONU:1824
:12,08772
:14,08627
Point de Fusion:0.0°C
:Boiling Pt:
:Boiling Pt: 1390 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 323 °C
:Flash Pt:
:Flash Pt:
:8
:III
Customers Who Bought This Also Bought