Nitrogen Standard, 1 ml = 1 mg N, 1000 ppm N as Ammonia (1216 ppm NH₃), Ricca Chemical Company
# de catalogue RC545516
Fournisseur: Ricca Chemical
Small Business Enterprise
Numéro CAS:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Concentration:1216/1000 ppm NH₃/N
- Packaging:Poly Bottle
- Description:Ammonia-Nitrogen standard solution
- Size:500 ml
- Cat. no.:RC545516
- Contents:NH₄Cl in H₂O
- Supplier No.:R5455000-500A
Spécifications
A propos de cet article
APHA for ammonia
1000 ppm nitrogen.
Customers Who Bought This Also Bought