Iodine monochloride solution acc. Wijs USP Test Solution (TS) for iodine absorption number of fats and oils
# de catalogue RC41101
Fournisseur: Ricca Chemical
Small Business Enterprise
Numéro CAS: 7790-99-0
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass, amber
- Size:4 l
- Cat. no.:RC41101
- Supplier No.:4110-1
Spécifications
Résultats des tests de spécification
- Appearance:Clear, dark brown, Acetic/Halogen odor
- Iodine/Chlorine Ratio:1.10 ± 0.05 Iodine/Chlorine Ratio
- Total Halogen Assay at 20 °C (traceable to NIST SRM 136):0.191 ± 0.002 N at 20.0 °C
Résultats des tests de spécification
A propos de cet article
Synonymes: Iodochloride
Formule:ClI
Numéro MDL:MFCD00011354
Numéro CAS:7790-99-0
:Iodine monochloride
:Storage Temperature: Ambient
Point d'ébullition:∼ 118 °C
Point de Fusion:26 °C
Densité:3.1 g/cm³ (29 °C)
ADR:8 (3),II
:Length
:Width
:Height
:Diameter
:Thickness
Point d'éclair:39 °C
:Boiling Pt:
:Boiling Pt: 97.3 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 26 °C
:Flash Pt:
:Flash Pt: 39 °C
:M.W. 162.36 g/mol
Index Merck:13,05039
Température d'entreposage:Ambient
EINECS:232-236-7
:97.3 °C (1013 hPa)
ONU:1792
:8
:II
:39 °C
:GHS05
:GHS07
:GHS02
:GHS06
Customers Who Bought This Also Bought