EDTA disodium salt 0.0100 M (M/100) in aqueous solution
# de catalogue RC270032
Fournisseur: Ricca Chemical
Small Business Enterprise
Numéro CAS: 139-33-3
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Poly bottle
- Size:32 oz.
- Cat. no.:RC270032
- Supplier No.:R2700000-1A
Spécifications
Résultats des tests de spécification
- Appearance:Clear, colorless, odorless
- Assay at 20 °C (traceable to NIST SRM 915):0.01000 ± 0.00001 M at 20.0 °C
Résultats des tests de spécification
A propos de cet article
Synonymes: Idranal III,Edetic acid disodium salt,Kestranal® 2S,Ethylenedinitrilotetra-acetic acid disodium salt,Disodium dihydrogen ethylenediaminetetraacetate
Formule:C₁₀H₁₄N₂Na₂O₈·2H₂O
Numéro MDL:MFCD00150037
Numéro CAS:6381-92-6
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Pt ébul.:|Point d'ébullition:
:Pt Fusion:|Point de fusion:
:Pt Eclair:|Point eclaire:
:M.W. 336,21 g/mol
:EDTA sel disodique
Point de Fusion:255 °C
Température d'entreposage:Température ambiante
Température d'entreposage:Ambient
EINECS:205-358-3
:12,03556
:Storage Temperature: Ambient
:Length
:Width
:Height
:Diameter
:Thickness
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 255 °C
:Flash Pt:
:Flash Pt:
:M.W. 372.24 g/mol
:EDTA disodium salt dihydrate
Customers Who Bought This Also Bought