Spécifications
- Conductivity:5000 µmhos/cm
- Size:946 ml (32 oz.)
- TDS:2620 ppm NaCl
- Supplier No.:R2245000-1A
Spécifications
A propos de cet article
Calibration standards are individually calibrated and certified directly traceable to National Institute of Standards and Technology Electrolytic Conductivity Standard Reference Materials.
Contact your VWR representative for custom-prepared conductivity standards.
Each plastic container is labeled with the certified conductivity value, TDS concentration expressed as ppm sodium chloride (NaCl), and TDS expressed as equivalent 'Fresh Water Ion' concentrations when appropriate.