Total Chlorine Accuracy Check Wastewater Standards, NSI
Catalogue # NSQCI-123
Fournisseur: SPEX CERTIPREP LLC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Numéro CAS:
Spécifications
- Concentration:1 mg/L (after dilution)
- Cat. no.:NSQCI-123
- Supplier No.:QCI-123
Spécifications
À propos de cet article
Standards are designed to verify the accuracy of residual chlorine analysis
Supplied in flame-sealed glass ampules.
Frequently Bought Together