Spécifications
- Description:Ready-To-Use
- Size:500 ml
- Supplier No.:QCI-071
Spécifications
A propos de cet article
Whole volume standard
Standard contains 500 mg/L of COD, 20 mg/L of NH3-N, 10 mg/L of N-NO3, 10 mg/L of p-PO4, 250 mg/L of SO4, and 161 mg/L of TOC.
Includes CoA listing true value and acceptance limits. Supplied in HDPE bottle.