Silicone Stoppers with Holes, Kimble Chase
# de catalogue KT953715-0801
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Cat. no.:KT953715-0801
- Stopper No.:8
- Hole Size:9.5 mm (3/8")
- Supplier No.:953715-0801
Spécifications
A propos de cet article
General-purpose, single-hole silicone rubber stoppers
Steam autoclavable. Opaque yellowish-white color. Stoppers are manufactured from pure silicone rubber.
Customers Who Bought This Also Bought