KIMBLE® Pressure Caps for NMR Sample Tubes, DWK Life Sciences
# de catalogue KT897095-0081
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Color:Yellow
- Description:PE Caps for 5 mm Tubes
- Cat. no.:KT897095-0081
- Supplier No.:897095-0081
Spécifications
A propos de cet article
Tight-fitting caps withstand vaporization pressure buildup within the sample tube.
Caps are precision machined and available in varied polyethylene sizes.
Ordering information: Pack of assorted 10 mm caps (KT897095-1011) contains 25 each of red, black, green, and yellow.
Customers Who Bought This Also Bought