Order Entry
ContactUsLinkComponent
Sample Tube Pressure Caps for 3, 5 and 10 mm Tubes
KIMBLE® Pressure Caps for NMR Sample Tubes, DWK Life Sciences
# de catalogue KT897095-0012
Sample Tube Pressure Caps for 3, 5 and 10 mm Tubes
KIMBLE® Pressure Caps for NMR Sample Tubes, DWK Life Sciences
# de catalogue KT897095-0012

Spécifications

  • Color:
    Assorted
  • Description:
    PE Caps for 5 mm Tubes
  • Cat. no.:
    KT897095-0012
  • Supplier No.:
    897095-0012

Spécifications

A propos de cet article

Tight-fitting caps withstand vaporization pressure buildup within the sample tube.

Caps are precision machined and available in varied polyethylene sizes.

Ordering information: Pack of assorted 10 mm caps (KT897095-1011) contains 25 each of red, black, green, and yellow.
Customers Who Bought This Also Bought