KONTES® Dounce Tissue Grinders, Kimble Chase
# de catalogue KT885300-0007
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Description:Small Pestle Clearance: 0.020 - 0.056 mm
- Cat. no.:KT885300-0007
- Tube Length:125 mm
- Supplier No.:885300-0007
Spécifications
A propos de cet article
For cellular work where the nucleus remains intact after homogenization
Borosilicate glass construction. One tube and two pestles are supplied with each tissue grinder. The large clearance pestle is used for initial sample reduction. The small clearance pestle forms the final homogenate.
Customers Who Bought This Also Bought