Rubber Sleeve, Plug-Type Stoppers, Kimble Chase
# de catalogue KT774261-0014
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Color:White
- Description:Plug-Type Stopper
- Cat. no.:KT774261-0014
- Fits Neck Size:14/20
- Bottom Diameter:14 mm
- Supplier No.:774261-0014
Spécifications
A propos de cet article
Hollow, plug-type stopper fits into the neck of the apparatus
Sleeve extension fits over the neck for a secure seal. Diaphragm can be punctured with a syringe needle. Use for outgassing NMR tubes, pressure venting or adding reactants via syringe.
Customers Who Bought This Also Bought