One-Way Stopcock Valves, Female to Male, Kimble Chase
# de catalogue KT420163-0000
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Color:Blue
- Description:One-way stopcock valve, polycarbonate body, HDPE plug, female luer to male luer
- Supplier No.:420163-0000
Spécifications
A propos de cet article
One-way stopcocks with a female to male connection and HDPE plug.
Customers Who Bought This Also Bought