Order Entry
Canada
ContactUsLinkComponent
HDPE Sterile Sample Bottles, Wide-Mouth, with Caps
I-CHEM™ Nalgene® High-Density Polyethylene Bottles, Sterile, Wide Mouth, Thermo Scientific
Catalog # IRN411-0125
Supplier:  Thermo Scientific
undefined
I-CHEM™ Nalgene® High-Density Polyethylene Bottles, Sterile, Wide Mouth, Thermo Scientific
Catalog # IRN411-0125
Supplier:  Thermo Scientific
Supplier Number:  N411-0125

Specifications

  • Cap Size:
    38-415
  • Capacity:
    125 ml (4.2 oz.)
  • Cat. no.:
    IRN411-0125
  • Supplier No.:
    N411-0125

Specifications

About this item

The I-CHEM Certified™ 400 Series bottles are processed to meet or exceed 'US EPA Specifications and Guidance for Contaminant-Free Sample Containers'. I-CHEM Certified™ 400 series bottles include a Certificate of Analysis with a bar-coded production number in every case

Bottles are gamma irradiated and certified for sterility in addition to metals and water quality parameters. Natural-colored bottles are made from high-density polyethylene and feature a wide-mouth and leakproof closure design. A heatshrink band on the neck and closure ensures sterility and provides added assurance that the closure is secure.

Each bottle in the 400 series has a bar-code label for absolute traceability.

Ordering information: All containers are supplied with caps attached.
Frequently Bought Together