Open-Top Septa Caps, Thermo Scientific
# de catalogue IR297-2400UB
Fournisseur: Thermo Scientific
UL Listed
Spécifications
- Cap Size:24-400
- Color:White
- Description:Open-Top Septa Cap
- Material:PP
- Liner Material:Bonded 0.060" PTFE-lined Silicone Septa
- Supplier No.:297-2400UB
Spécifications
A propos de cet article
These Open-Top Septa Caps are available with or without a bonded-in PTFE-lined silicone septa
They are the same high-quality polypropylene septa caps used on Thermo Scientific VOA vials, septa bottles, and septa jars. Caps are processed to meet EPA performance-based specifications for Volatile Organic Analysis.
Customers Who Bought This Also Bought