Order Entry
ContactUsLinkComponent
Boston Round Bottle, with Cap
I-CHEM™ Boston Round Bottles, with Caps, Thermo Scientific
# de catalogue IR249-1000
Fournisseur:  Thermo Scientific
Boston Round Bottle, with Cap
I-CHEM™ Boston Round Bottles, with Caps, Thermo Scientific
# de catalogue IR249-1000
Fournisseur:  Thermo Scientific

Spécifications

  • Cap Size:
    33-430
  • Capacity:
    1 L (33.8 oz.)
  • Color:
    Amber
  • Description:
    I-CHEM Certified™ 200 Series
  • Cat. no.:
    IR249-1000
  • Supplier No.:
    249-1000

Spécifications

A propos de cet article

Amber and clear glass bottles for use in the analysis of semivolatiles, pesticides, PCB’s, and metals.

  • Convenient, sturdy bottles for general sampling and use
  • Amber glass bottles are designed for light-sensitive samples
  • I-CHEM™ 100 Series bottles, for general use, are not processed or certified

I-CHEM Certified™ 200 and 300 series bottles are processed to meet or exceed 'US EPA Specifications and Guidance for Contaminent-Free Sample Containers' and include sample-analysis labels, unattached. I-CHEM Certified™ 300 series bottles include a Certificate of Analysis with a bar-coded production number in every case.

Each bottle in the 300 series has a manually readable bar-code lot label for absolute traceability and for use with automated sample tracking systems.

Ordering information: Available in amber and clear glass. Supplied with fluoropolymer resin-lined polypropylene caps attached.
Customers Who Bought This Also Bought