Spécifications
- Chemical method:Iron(III) thiocyanate
- Description:Hydrogen Peroxide CHEMets Refill
- Cat. no.:CXR-5510
- Range:0 - 0.8 and 1 - 10 ppm
- Supplier No.:R5510
Spécifications
A propos de cet article
Hydrogen Peroxide CHEMets® used to to monitor sterilisation solutions in the packaging and sanitizing processes.
Hydrogen peroxide is a strong oxidizing agent used for treating of industrial effluents and domestic waste and serving as a disinfectant in aseptic packaging.