Potassium dihydrogen phosphate ≥99.0%, crystals, Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
# de catalogue CAJT4008-5
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
Numéro CAS: 7778-77-0
MOC : With Information"Certificats
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle, amber, wide mouth
- Size:2.5 kg
- Supplier No.:4008-05
Spécifications
Résultats des tests de spécification
- For Laboratory,Research,or Manufacturing Use:
- Assay (KH₂PO₄):≥99.0 %
- DNase Activity (None Detected):Passes Test
- RNase Activity:Passes Test
- Protease Activity:Passes Test
- pH of 5% Solution at 25°C:4.1 - 4.5
- Insoluble Matter, Calcium and NH₄OH Precipitate:≤0.01 %
- Loss on Drying at 105°C:≤0.2 %
- Heavy Metals (as Pb):≤0.001 %
- Iron (Fe):≤0.002 %
- Trace Impurities - Arsenic (As):≤3.000 ppm
Résultats des tests de spécification
A propos de cet article
Synonymes: Monopotassiumphosphate,prim.-Potassium phosphate,Monopotassium phosphate,Potassium phosphate monobasic,Monobasic Potassium Phosphate
Point d'ébullition:∼100 °C (1013 hPa)
Point de Fusion:253 °C
Densité:2.3 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0
:Management of Change (MOC) category = NI
:Boiling Pt:
:Boiling Pt: ~ 100 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 253 °C
:Flash Pt:
:Flash Pt:
:M.W. 136.09 g/mol
Index Merck:13,07744
EINECS:231-913-4
:100
Température d'entreposage:Ambient
:01-2119490224-41
:GHS07
:M.O.C. Category NI
Customers Who Bought This Also Bought