di-Ammonium-fer (III) citrate (16,5 - 18,5% Fe) FCC
Glass bottle, amber, wide mouth, black T/E cap, 500 g
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Fournisseur #: BAKR1980-01
di-Ammonium-fer (III) citrate (16,5 - 18,5% Fe) FCC
Catalog # CAJT1980-1
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 1185-57-5
MOC: Réglementé LimitéCertificates
Jump to:
Product Details & Documents
Synonymes: Ammonium ferric citrate, Ferric Ammonium Citrate, Ammonium iron (III) citrate
- CAS Number:1185-57-5
- Formule:C₆H₅+4y)Fe(x)N(y)O₇
- Numéro MDL:MFCD00013099
- Ébullition Pt:197 °C (1013 hPa)
- Densité:1,8 g/cm³ (20 °C)
- :Management of Change (MOC) category = RL
- L'EINECS:214-686-6
Documents
Spécifications
Résultats des tests de spécification
- FCC - Assay (as Fe):16.5 - 18.5 %
- Identification:Passes Test
- Ferric Citrate:Passes Test
- Lead (Pb):≤2 mg/kg
- Mercury (Hg):≤1 mg/kg
- Oxalate (C₂O₄):Passes Test
- Sulfate (SO₄):≤0.3 %
- Tartrate:Passes Test
Spécifications
- Type de conditionnement:Glass bottle, amber, wide mouth, black T/E cap
- Taille:500 g
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



