Bulk Syringes, Nonsterile, BD Medical
10 ml, BD Luer-Lok®
Supplier: BD
: 301029
Jump to:
Product Details & Documents
Clean, ready-to-use, nonsterile syringes are siliconized and assembled with scale.
- Syringes can be autoclaved or gas sterilized out of poly bags
- Before autoclaving, remove plunger rod from barrel
Note: Syringes are supplied without needle. Needles must be purchased separately.
Ordering information: All syringes are bulk packed in double-wrapped poly bags. Sterile tip shield is sold separately.
Spécifications
- Volume:10 ml
- Tip Style:BD Luer-Lok®
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Sterile Tip Shield for Syringes; for use with BD Bulk Syringes, Nonsterile (301025 series)Sterile Tip Shield for Syringes; for use with BD Bulk Syringes, Nonsterile (301025 series)CABD305819Log in pour la disponibilité