Bulk Syringes, Nonsterile, BD Medical
10 ml, BD Luer-Lok®
Catalog # CABD301029
Supplier:   BD
 :  301029
Bulk Syringes, Nonsterile, BD Medical
Catalog # CABD301029
Supplier:  BD
Bulk Syringes, Nonsterile, BD Medical
Bulk Syringes, Nonsterile, BD Medical
10 ml, BD Luer-Lok®
Price
$611.00
Log in pour la disponibilité
Case of 850
Product Details & Documents
Clean, ready-to-use, nonsterile syringes are siliconized and assembled with scale.
  • Syringes can be autoclaved or gas sterilized out of poly bags
  • Before autoclaving, remove plunger rod from barrel
Note: Syringes are supplied without needle. Needles must be purchased separately.
Ordering information: All syringes are bulk packed in double-wrapped poly bags. Sterile tip shield is sold separately.
Spécifications
  • Volume:
    10 ml
  • Tip Style:
    BD Luer-Lok®
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Sterile Tip Shield for Syringes; for use with BD Bulk Syringes, Nonsterile (301025 series)
    Sterile Tip Shield for Syringes; for use with BD Bulk Syringes, Nonsterile (301025 series)
    CABD305819
    Log in pour la disponibilité
    Pack of 200 - $25.00