Acetone ≥99.5% (by GC, corrected for water content), GR ACS, Supelco®
# de catalogue CAAX0120-3
Fournisseur: MilliporeSigma
Numéro CAS: 67-64-1
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Steel barrel
- Size:20 L
- Cat. no.:CAAX0120-3
- Supplier No.:AX0120-3
Spécifications
Résultats des tests de spécification
- Assay (GC, corrected for water content):99.5% min
- Aldehyde (as HCHO):0.002% max
- Color (APHA):10 max
- Form:Clear liquid
- Isopropyl alcohol:0.05% max
- Methanol:0.05% max
- Residue after evaporation:0.001% max
- Solubility in water:To pass test
- Substances reducing permanganate:To pass test
- Titrable acid:0.0003meq/g max
- Titrable base:0.0006meq/g max
- Water (H2O):0.5% max
- Supelco®, Acetone ≥99.5% (by GC, corrected for water content), GR ACS:
Résultats des tests de spécification
A propos de cet article
Synonymes: 2-Propanone,DMK,Propanone
Point d'ébullition:56,2 °C (1013 hPa)
Point de Fusion:-95,4 °C
Densité:0,792 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
Point d'éclair:<–20 °C (closed cup)
:Point d'ébullition:
:Pt ébul.: 56,2 °C (1013 hPa)
:Point de fusion:
:Pt Fusion: -95,4 °C
:Point eclaire:
:Pt Eclair: Max. -20 °C (coupe fermée)
:M.W. 58,08 g/mol
Index Merck:13,00067
EINECS:200-662-2
:-95,4
:56,2
Température d'entreposage:Température ambiante
ONU:1090
:01-2119471330-49
:3
:II
:<–20 °C
:GHS02
:GHS07
Customers Who Bought This Also Bought