Spécifications
- Packaging:Powder Pillows
- Description:Chlorine, Total
- Cat. no.:CA97009-464
- Range:0 - 2.00 mg/L
- Supplier No.:2105669
Spécifications
A propos de cet article
This colorimeter is a portable, rugged, and convenient instrument, ideally suited for in-the-field analysis
Tough, impact-resistant shell is also waterproof and floats. Four buttons allow easy operation. Accuracy is comparable to expensive lab instruments. All units are factory-programmed; no manual calibration is ever required. All units come with enough reagent sets for 100 tests.