ContactUsLinkComponent
Dpd Chlorine Total Reagent Powder Pillows Pack Of 100
Pocket Colorimeter II Test Kits, Hach
# de catalogue CA97009-464
Fournisseur:  Hach
Numéro CAS:  
Pocket Colorimeter II Test Kits, Hach
# de catalogue CA97009-464
Fournisseur:  Hach
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Packaging:
    Powder Pillows
  • Description:
    Chlorine, Total
  • Cat. no.:
    CA97009-464
  • Range:
    0 - 2.00 mg/L
  • Supplier No.:
    2105669
Spécifications
A propos de cet article
This colorimeter is a portable, rugged, and convenient instrument, ideally suited for in-the-field analysis
Tough, impact-resistant shell is also waterproof and floats. Four buttons allow easy operation. Accuracy is comparable to expensive lab instruments. All units are factory-programmed; no manual calibration is ever required. All units come with enough reagent sets for 100 tests.
Customers Who Bought This Also Bought