ContactUsLinkComponent
High Range Test Strips
Chloride QuanTab® Test Strips, Hach
# de catalogue CA97009-462
Fournisseur:  Hach
Numéro CAS:  
Chloride QuanTab® Test Strips, Hach
# de catalogue CA97009-462
Fournisseur:  Hach
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Chemical method:
    Chloride
  • Cat. no.:
    CA97009-462
  • Range:
    300 - 6000 mg/L
  • Supplier No.:
    2751340
Spécifications
A propos de cet article
With Hach® water quality test strips, technicians in the field can test many samples in only a few minutes and make immediate evaluations on-site
These test strips quickly and quantitatively determine chloride levels in water samples. After dipping in the sample, the strip changes color and can be compared against an included scale for an accurate reading. Each pack contains 40 tests.
No measuring, set-up, clean-up, or chemical handling are necessary. Hach® test strips are also used in laboratories all over the world for pre-test screening to detect the presence of materials that might interfere with lab testing.
Customers Who Bought This Also Bought