ContactUsLinkComponent
SABARAUD DEXTROSE 500ML PK10
Sabouraud Dextrose Agar, BD
# de catalogue CA90005-224
Fournisseur:  BD
Numéro CAS:  
Sabouraud Dextrose Agar, BD
# de catalogue CA90005-224
Fournisseur:  BD
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Sabouraud Dextrose Agar, Sterile Pack
  • Size:
    500 mL
  • Cat. no.:
    CA90005-224
  • Supplier No.:
    299510
Spécifications
A propos de cet article
Sabouraud Dextrose Agar is used for the cultivation of dermatophytes.
  • Peptones provide a source of nitrogen
  • Dextrose is an energy source
  • Agar is the solidifying agent
After sufficient incubation, the plates should show isolated colonies in streaked areas and confluent growth in areas of heavy inoculation.
Customers Who Bought This Also Bought