ContactUsLinkComponent
Czapek Dox Broth
Czapek-Dox and Czapek Solution Media, BD Biosciences
# de catalogue CA90002-654
Fournisseur:  BD
Numéro CAS:  
Czapek-Dox and Czapek Solution Media, BD Biosciences
# de catalogue CA90002-654
Fournisseur:  BD
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Difco™ Czapek Dox Broth
  • Size:
    500 g
  • Cat. no.:
    CA90002-654
  • Supplier No.:
    233810
Spécifications
A propos de cet article
Czapek-Dox Broth and Czapek Solution Agar are used for cultivating fungi and bacteria capable of using inorganic nitrogen.
  • Saccharose is the sole carbon source
  • Sodium nitrate is the sole nitrogen source
  • Dipotassium phosphate is the buffering agent
  • Potassium chloride contains essential ions
  • Magnesium sulfate and ferrous sulfate are sources of cations
  • Agar is the solidifying agent in Czapek Solution Agar
Czapek-Dox media are useful in a variety of microbiological procedures, including soil microbiology and fungi and mildew resistance tests. These media produce moderately vigorous growth of most saprophytic aspergilli and yield characteristic mycelia and conidia.
Customers Who Bought This Also Bought