Spécifications
- Description:Difco™ Czapek Dox Broth
- Size:500 g
- Cat. no.:CA90002-654
- Supplier No.:233810
Spécifications
A propos de cet article
Czapek-Dox Broth and Czapek Solution Agar are used for cultivating fungi and bacteria capable of using inorganic nitrogen.
- Saccharose is the sole carbon source
- Sodium nitrate is the sole nitrogen source
- Dipotassium phosphate is the buffering agent
- Potassium chloride contains essential ions
- Magnesium sulfate and ferrous sulfate are sources of cations
- Agar is the solidifying agent in Czapek Solution Agar
Czapek-Dox media are useful in a variety of microbiological procedures, including soil microbiology and fungi and mildew resistance tests. These media produce moderately vigorous growth of most saprophytic aspergilli and yield characteristic mycelia and conidia.