ContactUsLinkComponent
CULTURESWAB EZ SWAB SNGL PK100
BD BBL™ CultureSwab® EZ Collection and Transport System, BD Diagnostics
# de catalogue CA90001-090
Fournisseur:  BD
CULTURESWAB EZ SWAB SNGL PK100
BD BBL™ CultureSwab® EZ Collection and Transport System, BD Diagnostics
# de catalogue CA90001-090
Fournisseur:  BD
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Single Swab Format
  • Cat. no.:
    CA90001-090
  • Supplier No.:
    220144
Spécifications
A propos de cet article
Medium-free systems prevent specimen dilution and contamination that can occur with devices containing transport media
Medium-free systems prevent specimen dilution and contamination that can occur with devices containing transport media
Systems feature swabs with a special polyurethane open-celled structure that releases organisms from their environment and protects them during transport. Swabs have a 13.3cm (51/4") long shaft to reach tough sampling sites. They fit into round-bottom transport tubes that stand upright in traditional racks for easy transportation. The caps for the transport tubes are color-coded for easy identification and the double action seal prevents dried out or leaky samples for increased safety.
Customers Who Bought This Also Bought