To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Depth:15 mm (19/32")
- Ø:95 mm (3 3/4")
- Description:Prostate Biopsy Container
- Cat. no.:CA89137-782
- Fill Type:Prefilled
- No. of Compartments:12
- Supplier No.:M970-D12P
Specifications
About this item
These polystyrene containers are half-filled with 10% neutral buffered formalin
The CoreDish® is available half prefilled with 10% neutral buffered formalin or empty for more versatility.
They feature leakproof closures with an O-ring, and clearly marked compartments. A label allows essential information to be written on the base. Different configurations are available for general purposes, breast, prostate, upper GI, and lower GI biopsies