Flanged Plug Caps, Simport Scientific
Catalog # CA60831-001
Supplier: Simport Scientific
Specifications
- Color:Blue
- Cat. no.:CA60831-001
- For Tubes:12 mm
- Supplier No.:T404-3B
Specifications
About this item
This line of polyethylene flanged plug caps provides exceptional leak-resistance in test tubes, centrifuge tubes, and round cuvettes.
Flanged plug caps feature two flexible flanges that press firmly against the inside wall of the tubes, thereby providing better leak-resistance than standard plug caps. Available in a variety of colors to facilitate identification.
Customers Who Bought This Also Bought