Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Falcon® Round-Bottom Tubes, Disposable, Polypropylene, Corning® | Avantor

Falcon® Round-Bottom Tubes, Disposable, Polypropylene, Corning®

5 ml, Plain Tube, without Cap, 75 mm, 12 mm, 125/Bag, Sterile
Catalog # CA60819-659
Supplier:   Corning
Fournisseur #:  352053
Falcon® Round-Bottom Tubes, Disposable, Polypropylene, Corning®
Catalog # CA60819-659
Supplier:  Corning
Falcon® Round-Bottom Tubes, Disposable, Polypropylene, Corning®
Falcon® Round-Bottom Tubes, Disposable, Polypropylene, Corning®

5 ml, Plain Tube, without Cap, 75 mm, 12 mm, 125/Bag, Sterile

Price
$454.00
Log in pour la disponibilité
Case of 1,000 (6ml)
Product Details & Documents

These disposable, clear polypropylene tubes are best suited for procedures requiring high optical clarity, and thermal and chemical stability. Provide reliable containment of laboratory fluid samples.

  • Dual position snap-cap prevents sample loss with ease of handling, heavier gauge walls, and unique construction provide a secure, positive seal
  • Medical-style packaging materials
  • 17×100 mm tubes have blue printed graduations and white writing patch
  • Tube dimensions and volumes are approximate
  • Widely used and referenced in laboratory protocols
  • Sterile, nonpyrogenicity tested to less than 0.1 EU/ml (Cat. No. 352002 nonsterile)

3000 RCF rating (RCF claims refer to relative centrifugal force measured in g-force for materials with a specific gravity of 1.0 used in an appropriate rotor with correct cushion and safety precautions. Tubes used with organic solvents at temperatures below 0 °C may have lower RCF ratings).

Spécifications
  • Ø:
    12 mm
  • Sterility:
    Sterile
  • Capacité:
    5 ml
  • Désignation:
    Plain Tube, without Cap
  • Hauteur:
    75 mm
  • Packaging:
    125/Bag
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.