Propeller Stirring Rod, METTLER TOLEDO®
# de catalogue CA46610-856
Fournisseur: Mettler Toledo
Spécifications
- Description:Propeller Stirring Rod
- Cat. no.:CA46610-856
- Supplier No.:101229
Spécifications
A propos de cet article
Propeller style stirring rod for use with DL50 Graphix and DL50 Rondolino Titrators.
Customers Who Bought This Also Bought