Spécifications
- Description:Donor Horse Serum, U.S. Sourced
- Size:500 mL
- Cat. no.:CA45001-058
- Supplier No.:35-030-CV
Spécifications
A propos de cet article
Horse serum is often used in testing media for mycoplasma contamination or as a blocking agent in immunoassays
- Donor Horse Serum is collected aseptically from a standing herd of healthy equines
- Collected from animals located entirely within the U.S.
- Donor animals are inspected periodically by a licensed veterinarian and are certified free of diseases of clawed animals
- For Research Use Only
It can also be combined with fetal bovine serum for the culture of rat adrenal pheochromocytoma (PC12) cells