ContactUsLinkComponent
DONOR HORSE SERUM EA=500ML
Corning® Donor Horse Serum, US Sourced, Corning
# de catalogue CA45001-058
Fournisseur:  Corning
DONOR HORSE SERUM EA=500ML
Corning® Donor Horse Serum, US Sourced, Corning
# de catalogue CA45001-058
Fournisseur:  Corning
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Donor Horse Serum, U.S. Sourced
  • Size:
    500 mL
  • Cat. no.:
    CA45001-058
  • Supplier No.:
    35-030-CV
Spécifications
A propos de cet article
Horse serum is often used in testing media for mycoplasma contamination or as a blocking agent in immunoassays
  • Donor Horse Serum is collected aseptically from a standing herd of healthy equines
  • Collected from animals located entirely within the U.S.
  • Donor animals are inspected periodically by a licensed veterinarian and are certified free of diseases of clawed animals
  • For Research Use Only
It can also be combined with fetal bovine serum for the culture of rat adrenal pheochromocytoma (PC12) cells
Customers Who Bought This Also Bought