Short-Term Detector Tubes, Draeger
# de catalogue CA13453-106
Fournisseur: Draeger
Spécifications
- Cat. no.:CA13453-106
- No. of Strokes:1
- Gas or Vapor Tube:Ammonia
- Standard Measurement Range:5 - 100 ppm
- Extended Measurement Range:—
- Supplier No.:8101941
Spécifications
A propos de cet article
For use with the accuro® Gas Detector Pump (see 34330-068 series) and Aerotest Breathing Air Tester (see 34330-078).
- Rapid and accurate readings
- Instantaneous measurement of hazardous substances
For rapid and accurate readings, these tubes serve the targeted measurement of instantaneous concentrations of hazardous substances and the short-term measurements are carried out manually with pump unit.
Ordering information: For additional tubes, contact your Avantor sales representative. Halogenated hydrocarbons detector tubes (13453-172) must be shipped via motor freight.
Customers Who Bought This Also Bought