Spécifications
- Clothing Size:3XL
- Garment application:Cleanroom,Laboratory
- Color:White
- Cat. no.:CA13451-042
- Chest Size:301/4"
- Inseam:311/2"
- Supplier No.:SL122TWH3X00
- Sleeve:381/2"
Spécifications
A propos de cet article
DuPont™ Tychem® 4000 delivers effective protection against a range of chemical environments.
DuPont™ Tychem® SL material provides effective protection against a wide range of chemicals. Tychem® SL material utilizes Saranex® 23-P film laminated to Tyvek® fabric, resulting in a rugged, durable, and abrasion-resistant garment. The fabric offers a minimum of 30 minutes of protection against over 124 chemical challenges. Coveralls are ideally suited for chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environment applications.
- Standard fit hood
- Elastic wrists
- Attached socks
- Storm flap with adhesive closure
- Taped seams
Tychem® 4000, utilizes Saranex™ 23-P film laminated to Tyvek® fabric to provide a rugged and durable fabric that resists abrasion. Tychem® 4000 fabric provides at least 30 minutes of protection against >124 chemical challenges. Typical applications: chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments. Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing. Coveralls feature an attached hood with elastic around face opening and elastic openings at wrists for a tighter fit. Storm flap covers zipper, which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper. Integrated socks are composed of the same garment material.
Tychem® 4000, utilizes Saranex™ 23-P film laminated to Tyvek® fabric to provide a rugged and durable fabric that resists abrasion. Tychem® 4000 fabric provides at least 30 minutes of protection against >124 chemical challenges. Typical applications: chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments. Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing. Coveralls feature an attached hood with elastic around face opening and elastic openings at wrists for a tighter fit. Storm flap covers zipper, which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper. Integrated socks are composed of the same garment material.