Specifications
- Pack type:Sachets (green)
- Size:30×20 ml
- Cat. no.:CA11238-440
- pH:7.00
- pH Accuracy:±0.02
- Supplier No.:51302047
Specifications
About this item
NIST-traceable buffer for pH electrode calibration and verification.
- NIST-traceable
- Ensure freshness
- Improve data
Also available in multiple packaging options.
Mettler Toledo buffer sachets ensure a fresh portion of buffer is used for each calibration, improving your pH data.