Rheodyne HPLC RheBuild kit valve repair accessory
# de catalogue 97058-330
Fournisseur: Rheodyne
Customers Who Bought This Also Bought
Hydrogen peroxide 30% stabilized ACS, VWR Chemicals BDH®
Fournisseur: VWR International
214,30 $ to 2 349,04 $
Potassium dihydrogen phosphate ≥99.0%, GR ACS, Supelco®
Fournisseur: MilliporeSigma
1 033,55 $ to 4 052,62 $
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
194,14 $ to 694,12 $