Inert Mobile Phase Bottle Cap, Microsolv
# de catalogue 97057-126
Fournisseur: MicroSolv Technology Corp
Spécifications
- Color:Blue
- Description:Bottle cap, medical grade PTFE insert with 3 holes
- Material:PE
- Thread Size:GL-45
- Supplier No.:58820-06
Spécifications
A propos de cet article
Most labs use this bottle cap to ensure that their mobile phase is not contaminated and thereby helps to comply with their HPLC GLP requirements
Your mobile phase will only contact medical grade PTFE so there will not be any contamination from plastic or wax film. Keeping these caps on the bottles with the tubing through holes prevents dust and dirt from getting in and minimizes outgassing or absorption of CO2 from the atmosphere. The cap assembly can be autoclaved.
Customers Who Bought This Also Bought