AQR™ Caps for RSA™ Autosampler Vials, MicroSolv Technology Corporation
Catalog # 97055-440
Supplier: MicroSolv Technology Corp
Specifications
- Cap size:9-425 mm
- Cap Type:Screw cap
- Septum:Pre-Slit, Bonded, Ultra Pure Clear Silicone/PTFE
- Material:PP
- Cat. no.:97055-440
- Cap Color:Light Blue
- Supplier No.:9509S-30C-B-M