ContactUsLinkComponent
RSA* Autosampler screw top Caps
AQR™ Caps for RSA™ Autosampler Vials, MicroSolv Technology Corporation
Catalog # 97055-440
RSA* Autosampler screw top Caps
AQR™ Caps for RSA™ Autosampler Vials, MicroSolv Technology Corporation
Catalog # 97055-440
Specifications
  • Cap size:
    9-425 mm
  • Cap Type:
    Screw cap
  • Septum:
    Pre-Slit, Bonded, Ultra Pure Clear Silicone/PTFE
  • Material:
    PP
  • Cat. no.:
    97055-440
  • Cap Color:
    Light Blue
  • Supplier No.:
    9509S-30C-B-M