ContactUsLinkComponent
RSA* Autosampler screw top Caps
AQR™ Caps for RSA™ Autosampler Vials, MicroSolv Technology Corporation
# de catalogue 97055-438
RSA* Autosampler screw top Caps
AQR™ Caps for RSA™ Autosampler Vials, MicroSolv Technology Corporation
# de catalogue 97055-438
Spécifications
  • Cap Size:
    9-425 mm
  • Cap Type:
    Screw cap
  • Septum:
    Pre-Slit, Bonded, Ultra Pure Clear Silicone/PTFE
  • Material:
    PP
  • Cap Color:
    Light Blue
  • Supplier No.:
    9509S-30C-B