AQR™ Caps for RSA™ Autosampler Vials, MicroSolv Technology Corporation
# de catalogue 97055-438
Fournisseur: MicroSolv Technology Corp
Spécifications
- Cap Size:9-425 mm
- Cap Type:Screw cap
- Septum:Pre-Slit, Bonded, Ultra Pure Clear Silicone/PTFE
- Material:PP
- Cap Color:Light Blue
- Supplier No.:9509S-30C-B